missing translation for 'onlineSavingsMsg'
Learn More

Methanol, 90% (v/v), Ricca Chemical

Catalog No. p-3251928
Change view
Click to view available options
Quantity:
1 L
4 L
Packaging:
Natural Poly Bottle
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
482332 1 L Natural Poly Bottle
48231 4 L Natural Poly Bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. 482332 Supplier Ricca Chemical Company Supplier No. 482332
Only null left

Chemical Identifiers

CAS 67-56-1 , 7732-18-5
Molecular Formula CH4O
Molecular Weight (g/mol) Mixture
MDL Number MFCD00004595
InChI Key OKKJLVBELUTLKV-UHFFFAOYSA-N
Synonym methyl alcohol, wood alcohol, carbinol, wood spirit, wood naphtha, methylol, methyl hydroxide, pyroxylic spirit, colonial spirit, columbian spirit
PubChem CID 887
ChEBI CHEBI:17790
IUPAC Name methanol
SMILES CO

Specifications

Color Colorless
CAS Min % 9.0, 89.0
CAS Max % 11.0, 91.0
Boiling Point 64.5°C
Quantity 1 L
Packaging Natural Poly Bottle
Physical Form Liquid
Percent Purity 90%
Grade Laboratory
Concentration or Composition (by Analyte or Components) 90% (v/v)
Chemical Name or Material Methanol
Show More Show Less