missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Methanol, 90% (v/v), Ricca Chemical
Supplier: Ricca Chemical Company RI48231
Specifications
| Methanol | |
| Colorless | |
| 11.0,91.0 | |
| 4 L | |
| Liquid | |
| MFCD00004595 | |
| OKKJLVBELUTLKV-UHFFFAOYSA-N | |
| methanol | |
| 887 | |
| 90% | |
| 90% (v/v) |
| 67-56-1,7732-18-5 | |
| 9.0,89.0 | |
| 64.5°C | |
| Natural Poly Bottle | |
| CH4O | |
| methyl alcohol, wood alcohol, carbinol, wood spirit, wood naphtha, methylol, methyl hydroxide, pyroxylic spirit, colonial spirit, columbian spirit | |
| CO | |
| Mixture | |
| CHEBI:17790 | |
| Laboratory |
Chemical Identifiers
| 67-56-1 | |
| Mixture | |
| OKKJLVBELUTLKV-UHFFFAOYSA-N | |
| 887 | |
| methanol |
| CH4O | |
| MFCD00004595 | |
| methyl alcohol, wood alcohol, carbinol, wood spirit, wood naphtha, methylol, methyl hydroxide, pyroxylic spirit, colonial spirit, columbian spirit | |
| CHEBI:17790 | |
| CO |