missing translation for 'onlineSavingsMsg'
Learn More

Methanol, 70% v/v, Reagents

Catalog No. p-200021594
Change view
Click to view available options
Quantity:
1 L
4 L
20 L
Packaging:
Cubitainer
Glass Bottle
Poly Bottle
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
01334751 1 L Poly Bottle
01334750 20 L Cubitainer
01334749 4 L Poly Bottle
01334748 4 L Glass Bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
Catalog No. 01334751 Supplier Labchem Inc Supplier No. CM1088001A
Only null left

Methanol, 70% v/v, Reagent, for general laboratory use. Manufactured in ISO 9001 facility.

Chemical Identifiers

CAS 67-56-1 , 7732-18-5
Molecular Formula CH4O
Molecular Weight (g/mol) 32.04
InChI Key OKKJLVBELUTLKV-UHFFFAOYSA-N
Synonym Methyl Alcohol, Wood Alcohol
IUPAC Name methanol
SMILES CO

Specifications

Color Colorless
Boiling Point 70°C
Quantity 1 L
Packaging Poly Bottle
Physical Form Liquid
Specific Gravity 0.8911
Percent Purity 70%
Grade Reagent
Chemical Name or Material Methanol, 70% v/v