missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Methanol, 70% v/v, Reagents
Methanol, 70% v/v, Reagent, for general laboratory use. Manufactured in ISO 9001 facility.
Supplier: Labchem Inc CM10880020F
Specifications
| Methanol, 70% v/v | |
| Colorless | |
| 20 L | |
| Liquid | |
| CH4O | |
| OKKJLVBELUTLKV-UHFFFAOYSA-N | |
| methanol | |
| 70% |
| 67-56-1,7732-18-5 | |
| 70°C | |
| Cubitainer | |
| 0.8911 | |
| Methyl Alcohol, Wood Alcohol | |
| CO | |
| 32.04 | |
| Reagent |
Chemical Identifiers
| 67-56-1 | |
| 32.04 | |
| Methyl Alcohol, Wood Alcohol | |
| CO |
| CH4O | |
| OKKJLVBELUTLKV-UHFFFAOYSA-N | |
| methanol |