missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Methanol, 50% v/v, Reagents
Methanol, 50% v/v, Reagent, for general laboratory use. Manufactured in ISO 9001 facility.
Supplier: Labchem Inc CM1227004C
Specifications
| Methanol, 50% v/v | |
| Colorless | |
| 4 L | |
| Liquid | |
| H2O | |
| XLYOFNOQVPJJNP-UHFFFAOYSA-N | |
| water | |
| 50% |
| 7732-18-5,67-56-1 | |
| 77°C | |
| Glass Bottle | |
| 0.928 | |
| Methyl Alcohol, Wood Alcohol | |
| O | |
| 18.02 | |
| Reagent |
Chemical Identifiers
| 7732-18-5 | |
| 18.02 | |
| Methyl Alcohol, Wood Alcohol | |
| O |
| H2O | |
| XLYOFNOQVPJJNP-UHFFFAOYSA-N | |
| water |