missing translation for 'onlineSavingsMsg'
Learn More

Methanol, 50% v/v, Reagents

Catalog No. 01334737
Change view
Click to view available options
Quantity:
1 L
4 L
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
01334737 1 L
01334736 4 L
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. 01334737 Supplier Labchem Inc Supplier No. CM1227001C

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Methanol, 50% v/v, Reagent, for general laboratory use. Manufactured in ISO 9001 facility.

Chemical Identifiers

CAS 7732-18-5 , 67-56-1
Molecular Formula H2O
Molecular Weight (g/mol) 18.02
InChI Key XLYOFNOQVPJJNP-UHFFFAOYSA-N
Synonym Methyl Alcohol, Wood Alcohol
IUPAC Name water
SMILES O

Specifications

Color Colorless
Quantity 1 L
Packaging Glass Bottle
Physical Form Liquid
Specific Gravity 0.928
Boiling Point 77°C
Percent Purity 50%
Grade Reagent
Chemical Name or Material Methanol, 50% v/v