missing translation for 'onlineSavingsMsg'
Learn More

Methanol, 50% (v/v), Ricca Chemical

Supplier:  Ricca Chemical Company 482032

 View more versions of this product

Catalog No. 4820-32


Only null left
Explore available promotions

Description

Description

Specifications

Specifications

Methanol
Colorless
51.0,51.0
1 L
Liquid
MFCD00004595
OKKJLVBELUTLKV-UHFFFAOYSA-N
methanol
887
50%
50% (v/v)
7732-18-5,67-56-1
49.0,49.0
66°C
Natural Poly Bottle
CH4O
methyl alcohol, wood alcohol, carbinol, wood spirit, wood naphtha, methylol, methyl hydroxide, pyroxylic spirit, colonial spirit, columbian spirit
CO
Mixture
CHEBI:17790
Laboratory

Chemical Identifiers

7732-18-5
Mixture
OKKJLVBELUTLKV-UHFFFAOYSA-N
887
methanol
CH4O
MFCD00004595
methyl alcohol, wood alcohol, carbinol, wood spirit, wood naphtha, methylol, methyl hydroxide, pyroxylic spirit, colonial spirit, columbian spirit
CHEBI:17790
CO
Product Suggestions

Product Suggestions

Videos
SDS Documents

Documents

Promotions

Promotions