missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Methanol, 50% (v/v), Ricca Chemical
Specifications
Specifications
| Color | Colorless |
| CAS Min % | 49.0 |
| CAS Max % | 51.0 |
| Quantity | 4 L |
| Packaging | Natural Poly Bottle |
| Physical Form | Liquid |
| Boiling Point | 66°C |
| Percent Purity | 50% |
| Grade | Laboratory |
| Concentration or Composition (by Analyte or Components) | 50% (v/v) |
| Show More |