missing translation for 'onlineSavingsMsg'
Learn More

Methanol, 10% w/v Made with HPLC grade components, Reagents

Catalog No. 01334810
Change view
Click to view available options
Quantity:
500 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
01334810 500 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 01334810 Supplier Labchem Inc Supplier No. CM002700500C

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Methanol, 10% w/v Made with HPLC grade components, Reagent, for general laboratory use. Manufactured in ISO 9001 facility.

Chemical Identifiers

CAS 7732-18-5 , 67-56-1
Molecular Formula H2O
Molecular Weight (g/mol) 18.02
InChI Key XLYOFNOQVPJJNP-UHFFFAOYSA-N
Synonym Methyl Alcohol, Wood Alcohol
IUPAC Name water
SMILES O

Specifications

Color Colorless
Quantity 500 mL
Packaging Glass Bottle
Physical Form Liquid
Boiling Point 93°C
Percent Purity 10% w/v
Grade Reagent
Chemical Name or Material Methanol, 10% w/v Made with HPLC grade components