missing translation for 'onlineSavingsMsg'
Learn More

Methanesulfonic Acid 99.0+%, TCI America™

Catalog No. p-7131983
Change view
Click to view available options
Quantity:
25 g
500 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
M009325G 25 g
M0093500G 500 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. M009325G Supplier TCI America Supplier No. M009325G

Chemical Identifiers

CAS 75-75-2
Molecular Formula CH4O3S
Molecular Weight (g/mol) 96.1
MDL Number MFCD00007518
InChI Key AFVFQIVMOAPDHO-UHFFFAOYSA-N
Synonym methylsulfonic acid, methanesulphonic acid, methanesulfonicacid, kyselina methansulfonova, methansulfonsaeure, ccris 2783, kyselina methansulfonova czech, ch3so3h, methane sulfonic acid, sulfomethane
PubChem CID 6395
ChEBI CHEBI:27376
IUPAC Name methanesulfonic acid
SMILES CS(=O)(=O)O

Specifications

Color Yellow
Boiling Point 122°C
Quantity 25 g
UN Number 2586
Formula Weight 96.10
Percent Purity ≥99.0% (T)
Physical Form Liquid
Chemical Name or Material Methanesulfonic Acid
EINECSNumber (2)-1582
RTECSNumber PB1140000
TSCA Yes