Learn More
Methanesulfonic acid, 98+%
CAS: 75-75-2 | CH4O3S | 96.1 g/mol
Supplier: Thermo Scientific Chemicals A135650E
Description
Methanesulfonic acid is used as a catalyst in organic reactions namely esterification, alkylation and condensation reactions due to its non- volatile nature and solubility in organic solvents. It is also involved in the production of starch esters, wax oxidate esters, benzoic acid esters, phenolic esters, or alkyl esters. It reacts with sodium borohydride in presence of polar solvent tetrahydrofuran to prepare borane-tetrahydrofuran complex. It finds application in batteries, because of its purity and chloride absence. In pharmaceutical industry, it is used for the manufacturing of active pharmaceutical ingredients like telmisartan and eprosartan. It is useful in ion chromatography and is a source of carbon and energy for some gram-negative methylotropic bacteria.It is involved in the deprotection of peptides.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
ApplicationsMethanesulfonic acid is used as a catalyst in organic reactions namely esterification, alkylation and condensation reactions due to its non- volatile nature and solubility in organic solvents. It is also involved in the production of starch esters, wax oxidate esters, benzoic acid esters, phenolic esters, or alkyl esters. It reacts with sodium borohydride in presence of polar solvent tetrahydrofuran to prepare borane-tetrahydrofuran complex. It finds application in batteries, because of its purity and chloride absence. In pharmaceutical industry, it is used for the manufacturing of active pharmaceutical ingredients like telmisartan and eprosartan. It is useful in ion chromatography and is a source of carbon and energy for some gram-negative methylotropic bacteria.It is involved in the deprotection of peptides.
Notes
Moisture, light and heat sensitive. Hygroscopic. Keep container tightly closed in a dry and well-ventilated place. Incompatible with amines, strong reducing agents, ethyl vinyl ether, hydrofluoric acid, strong oxidizing agents and bases.
Specifications
| Methanesulfonic acid | |
| 19°C to 20°C | |
| 166°C to 168°C (10 mmHg) | |
| CH4O3S | |
| CH3SO3H | |
| 2500 g | |
| 1446024 | |
| 14,5954 | |
| Miscible with water. Slightly miscible with benzene and toluene. Immiscible with paraffins. | |
| CS(=O)(=O)O | |
| 96.1 | |
| CHEBI:27376 | |
| ≥98% |
| 75-75-2 | |
| 1.481 | |
| 156°C (312°F) | |
| 1.43 | |
| MFCD00007518 | |
| UN2586 | |
| Hygroscopic | |
| methylsulfonic acid, methanesulphonic acid, methanesulfonicacid, kyselina methansulfonova, methansulfonsaeure, ccris 2783, kyselina methansulfonova czech, ch3so3h, methane sulfonic acid, sulfomethane | |
| AFVFQIVMOAPDHO-UHFFFAOYSA-N | |
| methanesulfonic acid | |
| 6395 | |
| 96.1 |
Chemical Identifiers
| 75-75-2 | |
| 96.1 | |
| AFVFQIVMOAPDHO-UHFFFAOYSA-N | |
| 6395 | |
| methanesulfonic acid |
| CH4O3S | |
| MFCD00007518 | |
| methylsulfonic acid, methanesulphonic acid, methanesulfonicacid, kyselina methansulfonova, methansulfonsaeure, ccris 2783, kyselina methansulfonova czech, ch3so3h, methane sulfonic acid, sulfomethane | |
| CHEBI:27376 | |
| CS(=O)(=O)O |
Safety and Handling
GHS H Statement
H314
Causes severe skin burns and eye damage.
P234-P260-P264b-P270-P271-P280-P303+P361+P353-P304+P340-P305+P351+P338-P310-P330-P331-P363-P390-P501c
H290-H302+H312-H314-H335
DOTInformation : Hazard Class: 8; Packaging Group: II
EINECSNumber : 200-898-6
RTECSNumber : PB1140000
TSCA : Yes
Recommended Storage : Ambient temperatures; Store under Nitrogen
RUO – Research Use Only