missing translation for 'onlineSavingsMsg'
Learn More

Methanesulfonic acid, 70% aq. soln.

Catalog No. p-7023841
Change view
Click to view available options
Quantity:
100 mL
500 mL
2500 mL
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAA17803AE 100 mL
AAA17803AP 500 mL
AAA178030F 2500 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. AAA17803AE Supplier Thermo Scientific Chemicals Supplier No. A17803AE

CAS: 75-75-2 | CH4O3S | 96.1 g/mol

Methane sulfonic acid 70% solution is used virtually in all electronic applications involving tin or tin-alloy plating from non fluoborate systems. It replaces phosphoric acid in detergent formulations, thereby helping to avoid environmental phosphate pollution. It is used in the preparation of polyaniline/graphene nanocomposites, which enhances its thermal and electrical properties. It is utilized for electropolishing of aluminum as well as in electroplating of noble metals like gold, platinum, palladium and their alloys.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Methane sulfonic acid 70% solution is used virtually in all electronic applications involving tin or tin-alloy plating from non fluoborate systems. It replaces phosphoric acid in detergent formulations, thereby helping to avoid environmental phosphate pollution. It is used in the preparation of polyaniline/graphene nanocomposites, which enhances its thermal and electrical properties. It is utilized for electropolishing of aluminum as well as in electroplating of noble metals like gold, platinum, palladium and their alloys.

Solubility
Miscible with water. Slightly miscible with benzene and toluene. Immiscible with paraffins.

Notes
Light and heat sensitive. Keep the container tightly closed in a dry and well-ventilated place. Incompatible with amines, strong reducing agents, ethyl vinyl ether, hydrofluoric acid, strong oxidizing agents and bases.

Chemical Identifiers

CAS 75-75-2
Molecular Formula CH4O3S
Molecular Weight (g/mol) 96.1
MDL Number MFCD00007518
InChI Key AFVFQIVMOAPDHO-UHFFFAOYSA-N
Synonym methylsulfonic acid, methanesulphonic acid, methanesulfonicacid, kyselina methansulfonova, methansulfonsaeure, ccris 2783, kyselina methansulfonova czech, ch3so3h, methane sulfonic acid, sulfomethane
PubChem CID 6395
ChEBI CHEBI:27376
IUPAC Name methanesulfonic acid
SMILES CS(=O)(=O)O

Specifications

Density 1.32
Flash Point >110°C (230°F)
Refractive Index 1.415
Quantity 100 mL
UN Number UN2586
Merck Index 14,5954
Solubility Information Miscible with water. Slightly miscible with benzene and toluene. Immiscible with paraffins.
Formula Weight 96.1
Concentration or Composition (by Analyte or Components) 70% aq. soln.
Chemical Name or Material Methanesulfonic acid
Hazard Category H290-H302+H312-H314-H335
Hazard Statement GHS H Statement
H314
Causes severe skin burns and eye damage.
Precautionary Statement P234-P260-P264b-P270-P271-P280-P303+P361+P353-P304+P340-P305+P351+P338-P310-P330-P331-P363-P390-P501c
DOTInformation Hazard Class: 8; Packaging Group: II
EINECSNumber 200-898-6
TSCA Yes
Recommended Storage Ambient temperatures

RUO – Research Use Only