missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Methanesulfonamide 98.0+%, TCI America™
Supplier: TCI America M0915100G
Specifications
| Methanesulfonamide | |
| 91°C | |
| MFCD00007940 | |
| methylsulfonamide, methane sulfonamide, methanesulphonamide, methansulfonamid, methyl sulfonamide, aminosulfane dioxide, methyl sulfonylamine, ch3so2nh2, mesylamine, methansulfonamide | |
| CS(N)(=O)=O | |
| 95.12 | |
| 95.12 | |
| Crystalline Powder |
| 3144-09-0 | |
| CH5NO2S | |
| 100 g | |
| HNQIVZYLYMDVSB-UHFFFAOYSA-N | |
| methanesulfonamide | |
| 72879 | |
| ≥98.0% (N) |
Chemical Identifiers
| 3144-09-0 | |
| 95.12 | |
| HNQIVZYLYMDVSB-UHFFFAOYSA-N | |
| 72879 | |
| CS(N)(=O)=O |
| CH5NO2S | |
| MFCD00007940 | |
| methylsulfonamide, methane sulfonamide, methanesulphonamide, methansulfonamid, methyl sulfonamide, aminosulfane dioxide, methyl sulfonylamine, ch3so2nh2, mesylamine, methansulfonamide | |
| methanesulfonamide |
Safety and Handling
TSCA : Yes