missing translation for 'onlineSavingsMsg'
Learn More

Methanesulfonamide 98.0+%, TCI America™

Catalog No. p-7132441
Change view
Click to view available options
Quantity:
25 g
100 g
500 g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
M091525G 25 g
M0915100G 100 g
M0915500G 500 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. M091525G Supplier TCI America Supplier No. M091525G
Only null left

Chemical Identifiers

CAS 3144-09-0
Molecular Formula CH5NO2S
Molecular Weight (g/mol) 95.12
MDL Number MFCD00007940
InChI Key HNQIVZYLYMDVSB-UHFFFAOYSA-N
Synonym methylsulfonamide, methane sulfonamide, methanesulphonamide, methansulfonamid, methyl sulfonamide, aminosulfane dioxide, methyl sulfonylamine, ch3so2nh2, mesylamine, methansulfonamide
PubChem CID 72879
IUPAC Name methanesulfonamide
SMILES CS(N)(=O)=O

Specifications

Melting Point 91°C
Quantity 25 g
Formula Weight 95.12
Percent Purity ≥98.0% (N)
Physical Form Crystalline Powder
Chemical Name or Material Methanesulfonamide