missing translation for 'onlineSavingsMsg'
Learn More

Methacycline hydrochloride Pharmaceutical Secondary Standard, MilliporeSigma™ Supelco™

Catalog No. 111012198
Change view
Click to view available options
Quantity:
1 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
111012198 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 111012198 Supplier Merck Emd Millipore Supplier No. PHR15941G

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Pharmaceutical secondary standards for application in quality control, provide pharma laboratories and manufacturers with a convenient and cost-effective alternative to the preparation of in-house working standards. Methacycline hydrochloride is one of the tetracycline antibiotics effective against a broad-spectrum of Gram-positive and Gram-negative bacteria. It is formed as an intermediate during the preparation of oxytetracycline. Its antibacterial activity is higher than that of tetracycline and oxytetracycline. It finds applications in human and veterinary medications as well as feed additives.

Specifications

CAS 3963-95-9
Purity Grade Notes These secondary standards offer multi-traceability to the USP, EP and BP primary standards, where they are available.
Linear Formula C22H22N2O8 · HCl
MDL Number MFCD04972012
Synonym Methacycline hydrochloride; 6-Demethyl-6-deoxy-6-methyleneoxytetracycline monohydrochloride
Type Certified Reference Material
Quantity 1 g
Formula Weight 478.88
Grade Certified Reference Material