Learn More
Thermo Scientific Chemicals Methacrylamide, 98%, extra pure
CAS: 79-39-0 | C4H7NO | 85.11 g/mol
Supplier: Thermo Scientific Chemicals 149205000
Description
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Specifications
| Methacrylamide | |
| 79-39-0 | |
| White to Yellow | |
| Authentic | |
| Glass bottle | |
| H2C=C(CH3)CONH2 | |
| 500 g | |
| methacrylamide, 2-methylacrylamide, methacrylic amide, methacrylic acid amide, 2-propenamide, 2-methyl, 2-methylpropenamide, methacryamide, methylacrylic amide, mhoromer bm801, alpha-methyl acrylic amide | |
| Solubility in water: 202g/L (20°C) | |
| CC(=C)C(=O)N | |
| 85.11 | |
| CHEBI:51759 | |
| 98% | |
| Crystals |
| 98% | |
| 108.0°C to 112.0°C | |
| 215.0°C | |
| 97.5% min. (GC) | |
| C4H7NO | |
| MFCD00008018 | |
| 02, II, 399 | |
| 0095.0°C | |
| FQPSGWSUVKBHSU-UHFFFAOYSA-N | |
| 2-methylprop-2-enamide | |
| 6595 | |
| 85.11 | |
| Extra Pure |
Chemical Identifiers
| 79-39-0 | |
| 85.11 | |
| FQPSGWSUVKBHSU-UHFFFAOYSA-N | |
| 6595 | |
| 2-methylprop-2-enamide |
| C4H7NO | |
| MFCD00008018 | |
| methacrylamide, 2-methylacrylamide, methacrylic amide, methacrylic acid amide, 2-propenamide, 2-methyl, 2-methylpropenamide, methacryamide, methylacrylic amide, mhoromer bm801, alpha-methyl acrylic amide | |
| CHEBI:51759 | |
| CC(=C)C(=O)N |
Safety and Handling
GHS H Statement
Harmful if swallowed.
Causes serious eye irritation.
GHS P Statement
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes.
Remove contact lenses,if present and easy to do.
Continue rinsing.
If exposed o
GHS Signal Word: Warning
EINECSNumber : 201-202-3
RTECSNumber : UC6475000
TSCA : TSCA
RUO – Research Use Only