missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific Chemicals Methacrylamide, 98%, extra pure

Catalog No. p-156307
Change view
Click to view available options
Quantity:
250 g
500 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AC149202500 250 g
AC149205000 500 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AC149202500 Supplier Thermo Scientific Chemicals Supplier No. 149202500
Only null left

CAS: 79-39-0 | C4H7NO | 85.11 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 79-39-0
Molecular Formula C4H7NO
Molecular Weight (g/mol) 85.11
MDL Number MFCD00008018
InChI Key FQPSGWSUVKBHSU-UHFFFAOYSA-N
Synonym methacrylamide, 2-methylacrylamide, methacrylic amide, methacrylic acid amide, 2-propenamide, 2-methyl, 2-methylpropenamide, methacryamide, methylacrylic amide, mhoromer bm801, alpha-methyl acrylic amide
PubChem CID 6595
ChEBI CHEBI:51759
IUPAC Name 2-methylprop-2-enamide
SMILES CC(=C)C(=O)N

Specifications

Melting Point 108.0°C to 112.0°C
Color White to Yellow
Boiling Point 215.0°C
Infrared Spectrum Authentic
Assay Percent Range 97.5% min. (GC)
Packaging Glass bottle
Linear Formula H2C=C(CH3)CONH2
Quantity 250 g
Beilstein 02, II, 399
Solubility Information Solubility in water: 202g/L (20°C)
Sublimation Point 0095.0°C
Formula Weight 85.11
Percent Purity 98%
Grade Extra Pure
Physical Form Crystals
Chemical Name or Material Methacrylamide, 98%
Show More Show Less
Hazard Category GHS Signal Word: Warning
Hazard Statement GHS H Statement
Harmful if swallowed.
Causes serious eye irritation.
Precautionary Statement GHS P Statement
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes.
Remove contact lenses,if present and easy to do.
Continue rinsing.
If exposed o
EINECSNumber 201-202-3
RTECSNumber UC6475000
TSCA TSCA

RUO – Research Use Only