missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Methacrolein (stabilized with HQ) 90.0+%, TCI America™
$138.61 - $1049.21
Chemical Identifiers
| CAS | 78-85-3 |
|---|---|
| Molecular Formula | C4H6O |
| Molecular Weight (g/mol) | 70.09 |
| MDL Number | MFCD00006974 |
| InChI Key | STNJBCKSHOAVAJ-UHFFFAOYSA-N |
| Synonym | methacrolein, methacrylaldehyde, 2-propenal, 2-methyl, isobutenal, 2-methylacrolein, 2-methylpropenal, 2-methyl-2-propenal, methylacrylaldehyde, 2-methylenepropanal, methacrylic aldehyde |
| PubChem CID | 6562 |
| IUPAC Name | 2-methylprop-2-enal |
| SMILES | CC(=C)C=O |
| Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
|---|---|---|---|---|---|---|---|---|---|
| Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
|
M007825ML
|
TCI America
M007825ML |
25 mL |
Each for $138.61
|
|
|||||
|
M0078100ML
|
TCI America
M0078100ML |
100 mL |
Each for $415.84
|
|
|||||
|
M0078500ML
|
TCI America
M0078500ML |
500 mL |
Each for $1,049.21
|
|
|||||
Chemical Identifiers
| 78-85-3 | |
| 70.09 | |
| STNJBCKSHOAVAJ-UHFFFAOYSA-N | |
| 6562 | |
| CC(=C)C=O |
| C4H6O | |
| MFCD00006974 | |
| methacrolein, methacrylaldehyde, 2-propenal, 2-methyl, isobutenal, 2-methylacrolein, 2-methylpropenal, 2-methyl-2-propenal, methylacrylaldehyde, 2-methylenepropanal, methacrylic aldehyde | |
| 2-methylprop-2-enal |
Specifications
| 78-85-3 | |
| Yellow | |
| C4H6O | |
| 25 mL | |
| methacrolein, methacrylaldehyde, 2-propenal, 2-methyl, isobutenal, 2-methylacrolein, 2-methylpropenal, 2-methyl-2-propenal, methylacrylaldehyde, 2-methylenepropanal, methacrylic aldehyde | |
| CC(=C)C=O | |
| 70.09 | |
| 70.09 | |
| Liquid |
| -81°C | |
| 68°C | |
| MFCD00006974 | |
| 2396 | |
| STNJBCKSHOAVAJ-UHFFFAOYSA-N | |
| 2-methylprop-2-enal | |
| 6562 | |
| ≥90.0% (GC) | |
| Methacrolein (stabilized with HQ) |
Safety and Handling
EINECSNumber : (2)-0522
RTECSNumber : OZ2625000
TSCA : Yes
Recommended Storage : Refrigerator