CAS RN 78-85-3
Methacrolein, 90%, stabilized
CAS: 78-85-3 Molecular Formula: C4H6O Molecular Weight (g/mol): 70.09 MDL Number: MFCD00006974 InChI Key: STNJBCKSHOAVAJ-UHFFFAOYSA-N Synonym: methacrolein,methacrylaldehyde,2-propenal, 2-methyl,isobutenal,2-methylacrolein,2-methylpropenal,2-methyl-2-propenal,methylacrylaldehyde,2-methylenepropanal,methacrylic aldehyde PubChem CID: 6562 IUPAC Name: 2-methylprop-2-enal SMILES: CC(=C)C=O
Methacrolein, 96%, stab. with hydroquinone
CAS: 78-85-3 Molecular Formula: C4H6O Molecular Weight (g/mol): 70.09 MDL Number: MFCD00006974 InChI Key: STNJBCKSHOAVAJ-UHFFFAOYSA-N Synonym: methacrolein,methacrylaldehyde,2-propenal, 2-methyl,isobutenal,2-methylacrolein,2-methylpropenal,2-methyl-2-propenal,methylacrylaldehyde,2-methylenepropanal,methacrylic aldehyde PubChem CID: 6562 IUPAC Name: 2-methylprop-2-enal SMILES: CC(=C)C=O
Methacrolein, 90%, stabilized, AcroSeal™, Thermo Scientific™
CAS: 78-85-3 Molecular Formula: C4H6O Molecular Weight (g/mol): 70.09 MDL Number: MFCD00006974 InChI Key: STNJBCKSHOAVAJ-UHFFFAOYSA-N Synonym: methacrolein,methacrylaldehyde,2-propenal, 2-methyl,isobutenal,2-methylacrolein,2-methylpropenal,2-methyl-2-propenal,methylacrylaldehyde,2-methylenepropanal,methacrylic aldehyde PubChem CID: 6562 IUPAC Name: 2-methylprop-2-enal SMILES: CC(=C)C=O
Methacrolein (stabilized with HQ) 90.0+%, TCI America™
CAS: 78-85-3 Molecular Formula: C4H6O Molecular Weight (g/mol): 70.09 MDL Number: MFCD00006974 InChI Key: STNJBCKSHOAVAJ-UHFFFAOYSA-N Synonym: methacrolein,methacrylaldehyde,2-propenal, 2-methyl,isobutenal,2-methylacrolein,2-methylpropenal,2-methyl-2-propenal,methylacrylaldehyde,2-methylenepropanal,methacrylic aldehyde PubChem CID: 6562 IUPAC Name: 2-methylprop-2-enal SMILES: CC(=C)C=O