missing translation for 'onlineSavingsMsg'
Learn More

Metformin Impurity F Pharmaceutical Secondary Standard, MilliporeSigma™ Supelco™

Catalog No. 111014373
Change view
Click to view available options
Quantity:
200 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
111014373 200 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 111014373 Supplier Merck Emd Millipore Supplier No. PHR1532200MG

Pharmaceutical secondary standards for application in quality control, provide pharma laboratories and manufacturers with a convenient and cost-effective alternative to the preparation of in-house working standards.

Specifications

CAS 506-59-2
Purity Grade Notes These secondary standards offer multi-traceability to the USP, EP and BP primary standards, where they are available.
Linear Formula (CH3)2 NH · HCl
MDL Number MFCD00012477
Melting Point 170°C to 173°C
Synonym Dimethylamine hydrochloride; Dimethylamine Hydrochloride
Type Certified Reference Material
Quantity 200 mg
Formula Weight 81.54
Grade Certified Reference Material