missing translation for 'onlineSavingsMsg'
Learn More

Metformin hydrochloride Pharmaceutical Secondary Standard, MilliporeSigma™ Supelco™

Catalog No. 111012473
Change view
Click to view available options
Quantity:
500 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
111012473 500 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 111012473 Supplier Merck Emd Millipore Supplier No. PHR1084500MG

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Metformin is a biguanide hypoglycaemic agent and an oral antidiabetic medication widely used in the treatment of type 2 diabetes. It is effective in glycemic control and decreases the intestinal absorption of glucose, without the problem of serious lactic acidosis.

Specifications

CAS 1115-70-4
Purity Grade Notes These secondary standards offer multi-traceability to the USP, EP (PhEur) and BP primary standards, where they are available.
Physical Form Neat
Linear Formula NH2 C(=NH)NHC(=NH)N(CH3)2 · HCl
MDL Number MFCD00012582
Melting Point 223°C to 226°C
Synonym 1,1-Dimethylbiguanide hydrochloride; Metformin
Type Certified Reference Material
Quantity 500 mg
Formula Weight 165.62
Grade Certified Reference Material
Show More Show Less