missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Metformin-(dimethyl-d6) hydrochloride, ≥95.0% (HPLC), MilliporeSigma™ Supelco™
Specifications
Specifications
| CAS | 1185166-01-1 |
| Physical Form | Neat |
| Linear Formula | C4D6H5N5 · HCl |
| MDL Number | MFCD06658649 |
| Synonym | 1,1-Di(methyl-d3)biguanide hydrochloride; [2H6 ]-Metformin hydrochloride |
| Type | Analytical Standard |
| Quantity | 5 mg |
| Formula Weight | 171.66 |
| Percent Purity | ≥95.0% (HPLC) |
| Grade | Analytical Standard |