missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Metaphosphoric Acid, 3% w/v, Reagents
Metaphosphoric Acid, 3% w/v, Reagent, for general laboratory use. Manufactured in ISO 9001 facility.
Supplier: Labchem Inc CM10350020F
Specifications
| Metaphosphoric Acid, 3% w/v | |
| 7732-18-5,64-19-7 | |
| Cubitainer | |
| 20 L | |
| Liquid | |
| XLYOFNOQVPJJNP-UHFFFAOYSA-N | |
| water | |
| 3% |
| Colorless | |
| 100°C | |
| 1.019 | |
| H2O | |
| Phosphenic Acid | |
| O | |
| 18.02 | |
| Reagent |
Chemical Identifiers
| 7732-18-5 | |
| 18.02 | |
| Phosphenic Acid | |
| O |
| H2O | |
| XLYOFNOQVPJJNP-UHFFFAOYSA-N | |
| water |