missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ METAP2 Recombinant Rabbit Monoclonal Antibody (23GB2565)

Rabbit Recombinant Monoclonal Antibody
Supplier: Invitrogen™ MA553516
Description
METAP2 Recombinant Monoclonal Antibody for Flow, ICC/IF, Western Blot
The ADP-ribosylation factor (Arf) family are highly conserved members of the Ras superfamily of regulatory GTP-binding proteins. Arf proteins participate in routing of intracellular proteins to and within the Golgi complex. Cellular functions include maintenance of organelle integrity, coat protein assembly, as an activator of phospholipase D. The Arf family is divided functionally into the Arf and the Arf-like (Arl) proteins. The ARF proteins are categorized as class I (ARF1, ARF2 and ARF3), class II (ARF4 and ARF5) and class III (ARF6) and members of each class share a common gene organization.Chemical Identifiers
| METAP2 | |
| 0.4 mg/mL | |
| Flow Cytometry, Western Blot, Immunocytochemistry | |
| Unconjugated | |
| Rabbit | |
| RUO | |
| PBS with 50% glycerol and 0.02% sodium azide; pH 7.4 | |
| O08663, P38062, P50579 | |
| 10988, 56307, 64370 | |
| A synthesized peptide derived from human Methionine Aminopeptidase 2 (420-478AA). | |
| Antibody |
| 23GB2565 | |
| -20°C | |
| Recombinant Monoclonal | |
| Liquid | |
| IgG | |
| Human, Mouse, Rat | |
| METAP2 | |
| 4930584B20Rik; A930035J23Rik; AI047573; AL024412; Amp2; AU014659; eIF-2-associated p67; eIF-2-associated p67 homolog; initiation factor 2 associated 67 kDa protein; initiation factor 2-associated 67 kDa glycoprotein; initiation factor 2-associated protein (p67); MAP 2; MAP2; metAP 2; Metap2; Methionine aminopeptidase 2; methionyl aminopeptidase 2; Mnpep; p67; p67eIF2; Peptidase M; peptidase M 2; similar to methionyl aminopeptidase 2; testicular tissue protein Li 17 | |
| METAP2 | |
| Primary | |
| Affinity chromatography |
Specifications
| METAP2 | |
| Recombinant Monoclonal | |
| 0.4 mg/mL | |
| PBS with 50% glycerol and 0.02% sodium azide; pH 7.4 | |
| O08663, P38062, P50579 | |
| METAP2 | |
| A synthesized peptide derived from human Methionine Aminopeptidase 2 (420-478AA). | |
| 100 μL | |
| Primary | |
| Human, Mouse, Rat | |
| Antibody | |
| IgG |
| Flow Cytometry, Western Blot, Immunocytochemistry | |
| 23GB2565 | |
| Unconjugated | |
| METAP2 | |
| 4930584B20Rik; A930035J23Rik; AI047573; AL024412; Amp2; AU014659; eIF-2-associated p67; eIF-2-associated p67 homolog; initiation factor 2 associated 67 kDa protein; initiation factor 2-associated 67 kDa glycoprotein; initiation factor 2-associated protein (p67); MAP 2; MAP2; metAP 2; Metap2; Methionine aminopeptidase 2; methionyl aminopeptidase 2; Mnpep; p67; p67eIF2; Peptidase M; peptidase M 2; similar to methionyl aminopeptidase 2; testicular tissue protein Li 17 | |
| Rabbit | |
| Affinity chromatography | |
| RUO | |
| 10988, 56307, 64370 | |
| -20°C | |
| Liquid |