missing translation for 'onlineSavingsMsg'
Learn More

Metanilic acid, 98%, Thermo Scientific™

Supplier:  Thermo Scientific Chemicals 413710010

Quantity 1kg
Packaging Glass Bottle

 View more versions of this product

Catalog No. AC413710010

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.


Description

Description

This Thermo Scientific brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

Specifications

Chemical Identifiers

121-47-1
173.19
ZAJAQTYSTDTMCU-UHFFFAOYSA-N
8474
3-aminobenzenesulfonic acid
C6H7NO3S
MFCD00065345
metanilic acid, m-sulfanilic acid, m-aminobenzenesulfonic acid, m-anilinesulfonic acid, 1-aminobenzene-3-sulfonic acid, benzenesulfonic acid, 3-amino, aminobenzenesulfonic acid, kyselina metanilova, 3-amino-benzenesulfonic acid, 3-sulfoaniline
CHEBI:27764
C1=CC(=CC(=C1)S(=O)(=O)O)N

Specifications

Metanilic acid
121-47-1
98%
C6H7NO3S
1kg
15, 5998
Solubility in water: slightly soluble
C1=CC(=CC(=C1)S(=O)(=O)O)N
173.19
CHEBI:27764
98%
98%
Authentic
Glass Bottle
NH2C6H4SO3H
MFCD00065345
metanilic acid, m-sulfanilic acid, m-aminobenzenesulfonic acid, m-anilinesulfonic acid, 1-aminobenzene-3-sulfonic acid, benzenesulfonic acid, 3-amino, aminobenzenesulfonic acid, kyselina metanilova, 3-amino-benzenesulfonic acid, 3-sulfoaniline
ZAJAQTYSTDTMCU-UHFFFAOYSA-N
3-aminobenzenesulfonic acid
8474
173.19
Product Suggestions

Product Suggestions

Videos
Safety and Handling

Safety and Handling

GHS H Statement:
Harmful if swallowed.
Harmful in contact with skin.
Harmful if inhaled.

GHS P Statement:
Wear protective gloves/protective clothing/eye protection/face protection.
IF SWALLOWED: Immediately call a POISON CENTER or doctor/physician.
IF ON SKIN: Wash with plenty of soap and water.
IF INHALED: Remove victim to fresh air and keep at rest in a position comfortable for breathing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

GHS Signal Word: Warning

EINECSNumber : 204-473-6

RTECSNumber : OY2300000

TSCA : TSCA

SDS
Documents

Documents

Promotions

Promotions

Promo
Expires: 06-30-2027
Get Up to $1,800 in Chemicals with Purchase
Spend $250 or more on Thermo Fisher Scientific chemicals and get up to $1,800 to spend on additional chemicals.

Adding...