missing translation for 'onlineSavingsMsg'
Learn More

Metanilic acid, 98%, Thermo Scientific™

Catalog No. p-156292
Change view
Click to view available options
Quantity:
100g
250g
1kg
Packaging:
Glass bottle
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
AC413711000 100g Glass bottle
AC413712500 250g Glass bottle
AC413710010 1kg Glass bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. AC413711000 Supplier Thermo Scientific Chemicals Supplier No. 413711000

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

This Thermo Scientific brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

Chemical Identifiers

CAS 121-47-1
Molecular Formula C6H7NO3S
Molecular Weight (g/mol) 173.19
MDL Number MFCD00065345
InChI Key ZAJAQTYSTDTMCU-UHFFFAOYSA-N
Synonym metanilic acid, m-sulfanilic acid, m-aminobenzenesulfonic acid, m-anilinesulfonic acid, 1-aminobenzene-3-sulfonic acid, benzenesulfonic acid, 3-amino, aminobenzenesulfonic acid, kyselina metanilova, 3-amino-benzenesulfonic acid, 3-sulfoaniline
PubChem CID 8474
ChEBI CHEBI:27764
IUPAC Name 3-aminobenzenesulfonic acid
SMILES C1=CC(=CC(=C1)S(=O)(=O)O)N

Specifications

Infrared Spectrum Authentic
Assay Percent Range 98%
Packaging Glass bottle
Linear Formula NH2C6H4SO3H
Quantity 100g
Merck Index 15, 5998
Solubility Information Solubility in water: slightly soluble
Formula Weight 173.19
Percent Purity 98%
Chemical Name or Material Metanilic acid, 98%
Hazard Category GHS Signal Word: Warning
Hazard Statement GHS H Statement:
Harmful if swallowed.
Harmful in contact with skin.
Harmful if inhaled.
Precautionary Statement GHS P Statement:
Wear protective gloves/protective clothing/eye protection/face protection.
IF SWALLOWED: Immediately call a POISON CENTER or doctor/physician.
IF ON SKIN: Wash with plenty of soap and water.
IF INHALED: Remove victim to fresh air and keep at rest in a position comfortable for breathing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.
EINECSNumber 204-473-6
RTECSNumber OY2300000
TSCA TSCA

RUO – Research Use Only