missing translation for 'onlineSavingsMsg'
Learn More

Metanil Yellow, ≥98.0% (HPLC), MilliporeSigma™ Supelco™

Catalog No. 111014766
Change view
Click to view available options
Quantity:
100 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
111014766 100 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 111014766 Supplier Merck Emd Millipore Supplier No. 44426100MG
Only null left

Metanil Yellow (MY) belongs to the azo group of dyes.

Cleaning Products, Cosmetics, Food and Beverages and Personal Care

Specifications

Packaging Bottomless Glass Bottle with Inserted Fused Cone
CAS 587-98-4
Physical Form Neat
Linear Formula C18H14N3 NaO3 S
MDL Number MFCD00007486
Synonym 3-(4-Anilinophenylazo)benzenesulfonic acid sodium salt; Acid Yellow 36
Type Analytical Standard
Quantity 100 mg
Formula Weight 375.38
Percent Purity ≥98.0% (HPLC)
Grade Analytical Standard
Show More Show Less