Learn More
Thermo Scientific Chemicals Metanil Yellow
CAS: 587-98-4 | C18H14N3NaO3S | 375.378 g/mol
Supplier: Thermo Scientific Chemicals 413720010
Description
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Specifications
| Metanil Yellow | |
| 88% min. | |
| Acid Yellow 36, 3-[[4-(Phenylamino)phenyl]azo]benzenesulfonic acid monosodium salt, C.I. 13065, 3-(4-Anilinophenylazo)benzenesulfonic acid sodium salt | |
| NYGZLYXAPMMJTE-UHFFFAOYSA-M | |
| C1=CC=C(C=C1)NC2=CC=C(C=C2)N=NC3=CC(=CC=C3)S(=O)(=O)[O-].[Na+] | |
| 375.378 | |
| CHEBI:87235 | |
| 375.38 | |
| 1 kg |
| 587-98-4 | |
| C18H14N3NaO3S | |
| Solubility in water: 25g/L (20°C). Other solubilities: soluble in alcohol, moderately soluble in benzene and ether, slightly soluble in acetone | |
| Authentic | |
| sodium;3-[(4-anilinophenyl)diazenyl]benzenesulfonate | |
| 3935589 | |
| 413 to 419nm (in methanol) | |
| Glass bottle |
Chemical Identifiers
| 587-98-4 | |
| 375.378 | |
| Acid Yellow 36, 3-[[4-(Phenylamino)phenyl]azo]benzenesulfonic acid monosodium salt, C.I. 13065, 3-(4-Anilinophenylazo)benzenesulfonic acid sodium salt | |
| CHEBI:87235 | |
| C1=CC=C(C=C1)NC2=CC=C(C=C2)N=NC3=CC(=CC=C3)S(=O)(=O)[O-].[Na+] |
| C18H14N3NaO3S | |
| NYGZLYXAPMMJTE-UHFFFAOYSA-M | |
| 3935589 | |
| sodium;3-[(4-anilinophenyl)diazenyl]benzenesulfonate |
Safety and Handling
GHS H Statement Causes serious eye damage. Toxic to aquatic life with long lasting effects.
GHS P Statement IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing. Immediately call a POISON CENTER or doctor/physician. Avoid release to the environment.
Danger
RUO – Research Use Only