accessibility menu, dialog, popup

UserName

HNNNSOOONa(E/Z)

CAS RN 587-98-4

IUPAC Name: sodium 3-{2-[4-(phenylamino)phenyl]diazen-1-yl}benzene-1-sulfonate
Synonyms: colouring agent CI 13065
Molecular Weight (g/mol): 375.38
Molecular Formula: C18H14N3NaO3S
InChi Key: NYGZLYXAPMMJTE-UHFFFAOYSA-M
SMILES: [Na+].[O-]S(=O)(=O)C1=CC=CC(=C1)N=NC1=CC=C(NC2=CC=CC=C2)C=C1
Quantity 
  • (1)
  • (1)
  • (1)
  • (1)
pH 
  • (2)
Color 
  • (1)
Grade 
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (3)
  • (1)
  • (3)
  • (1)
  • (1)
  • (1)
  • (1)
  • (2)
  • (1)
  • (1)