missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Metamitron 98.0+%, TCI America™
$39.14 - $109.98
Chemical Identifiers
| CAS | 41394-05-2 |
|---|---|
| Molecular Formula | C10H10N4O |
| Molecular Weight (g/mol) | 202.217 |
| MDL Number | MFCD00055524 |
| InChI Key | VHCNQEUWZYOAEV-UHFFFAOYSA-N |
| Synonym | metamitron, goltix, herbrak, metamiton, 4-amino-3-methyl-6-phenyl-1,2,4-triazin-5 4h-one, metamitron german, methiamitron, methiamitron french, methiamitron belgium, bay-drw 1139 |
| PubChem CID | 38854 |
| ChEBI | CHEBI:6791 |
| IUPAC Name | 4-amino-3-methyl-6-phenyl-1,2,4-triazin-5-one |
| SMILES | CC1=NN=C(C(=O)N1N)C2=CC=CC=C2 |
Chemical Identifiers
| 41394-05-2 | |
| 202.217 | |
| VHCNQEUWZYOAEV-UHFFFAOYSA-N | |
| 38854 | |
| 4-amino-3-methyl-6-phenyl-1,2,4-triazin-5-one |
| C10H10N4O | |
| MFCD00055524 | |
| metamitron, goltix, herbrak, metamiton, 4-amino-3-methyl-6-phenyl-1,2,4-triazin-5 4h-one, metamitron german, methiamitron, methiamitron french, methiamitron belgium, bay-drw 1139 | |
| CHEBI:6791 | |
| CC1=NN=C(C(=O)N1N)C2=CC=CC=C2 |
Specifications
| 41394-05-2 | |
| White-Yellow | |
| MFCD00055524 | |
| 3077 | |
| VHCNQEUWZYOAEV-UHFFFAOYSA-N | |
| 4-amino-3-methyl-6-phenyl-1,2,4-triazin-5-one | |
| 38854 | |
| 202.22 | |
| Crystalline Powder |
| 168°C | |
| C10H10N4O | |
| 5 g | |
| metamitron, goltix, herbrak, metamiton, 4-amino-3-methyl-6-phenyl-1,2,4-triazin-5 4h-one, metamitron german, methiamitron, methiamitron french, methiamitron belgium, bay-drw 1139 | |
| CC1=NN=C(C(=O)N1N)C2=CC=CC=C2 | |
| 202.217 | |
| CHEBI:6791 | |
| ≥98.0% (GC,T) | |
| Metamitron |
Safety and Handling
RTECSNumber : XZ3015000
TSCA : No