missing translation for 'onlineSavingsMsg'
Learn More

Metamitron 98.0+%, TCI America™

Catalog No. M23885G
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
M23885G 5 g
M238825G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. M23885G Supplier TCI America Supplier No. M23885G
Only null left

Chemical Identifiers

CAS 41394-05-2
Molecular Formula C10H10N4O
Molecular Weight (g/mol) 202.217
MDL Number MFCD00055524
InChI Key VHCNQEUWZYOAEV-UHFFFAOYSA-N
Synonym metamitron, goltix, herbrak, metamiton, 4-amino-3-methyl-6-phenyl-1,2,4-triazin-5 4h-one, metamitron german, methiamitron, methiamitron french, methiamitron belgium, bay-drw 1139
PubChem CID 38854
ChEBI CHEBI:6791
IUPAC Name 4-amino-3-methyl-6-phenyl-1,2,4-triazin-5-one
SMILES CC1=NN=C(C(=O)N1N)C2=CC=CC=C2

Specifications

Melting Point 168°C
Color White-Yellow
Quantity 5 g
UN Number 3077
Formula Weight 202.22
Percent Purity ≥98.0% (GC,T)
Physical Form Crystalline Powder
Chemical Name or Material Metamitron
RTECSNumber XZ3015000
TSCA No