missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ Metadherin Polyclonal Antibody
GREENER_CHOICE

Catalog No. FEPA5144202
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
FEPA5144202 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. FEPA5144202 Supplier Invitrogen™ Supplier No. PA5144202
Only null left

Rabbit Polyclonal Antibody

Metadherin Polyclonal Antibody for IHC (P), IP, ELISA

Metadherin (Metastasis adhesion protein), also known as MTDH, LYsine-RIch CEACAM1 co-isolated (LYRIC), is a novel protein that localizes with the tight junction proteins ZO-1 and occludin in polarized epithelial cells. At the tight junction, it acts not as a structural component, but is rather recruited during the maturation of the tight junction complex. Metadherin is overexpressed in breast cancer tissue and breast tumor xenografts, while much lower levels are expressed in normal breast tissue. Metadherin binds to lung vasculature, one of the four common sites of breast cancer metastasis, through a C-terminal segment in the extracellular domain; blocking this lung-homing domain with antibodies or inhibiting metadherin with siRNA has been reported to inhibit breast cancer metastasis.

Specifications

Antigen Metadherin
Applications ELISA, Immunohistochemistry (Paraffin), Immunoprecipitation
Classification Polyclonal
Concentration 0.95 mg/mL
Conjugate Unconjugated
Formulation PBS with 50% glycerol and 0.02% sodium azide; pH 7.3
Gene MTDH
Gene Accession No. Q86UE4
Gene Alias 2610103J23Rik; 3D3; 3D3/Lyric; AEG1; AEG-1; astrocyte elevated gene 1; astrocyte elevated gene-1 protein; AV353288; D8Bwg1112e; LYRIC; LYRIC/ MTDH; LYRIC/3D3; lysine-rich CEACAM1 co-isolated protein; Metadherin; metastasis adhesion protein; MTDH; Protein LYRIC
Gene Symbols MTDH
Host Species Rabbit
Immunogen Recombinant Human Protein LYRIC protein (1-260AA).
Purification Method Antigen affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 92140
Target Species Human
Content And Storage -20°C or -80°C if preferred
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less