missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Metacresol Purple, TCI America™
$67.38 - $660.33
Chemical Identifiers
| CAS | 2303-01-7 |
|---|---|
| Molecular Formula | C21H18O5S |
| Molecular Weight (g/mol) | 382.43 |
| MDL Number | MFCD00005871 |
| InChI Key | OLQIKGSZDTXODA-UHFFFAOYSA-N |
| Synonym | m-Cresol Purple |
| PubChem CID | 73030 |
| IUPAC Name | 4-[3-(4-hydroxy-2-methylphenyl)-1,1-dioxo-2,1$l^{6}-benzoxathiol-3-yl]-3-methylphenol |
| SMILES | CC1=C(C=CC(=C1)O)C2(C3=CC=CC=C3S(=O)(=O)O2)C4=C(C=C(C=C4)O)C |
Chemical Identifiers
| 2303-01-7 | |
| 382.43 | |
| OLQIKGSZDTXODA-UHFFFAOYSA-N | |
| 73030 | |
| CC1=C(C=CC(=C1)O)C2(C3=CC=CC=C3S(=O)(=O)O2)C4=C(C=C(C=C4)O)C |
| C21H18O5S | |
| MFCD00005871 | |
| m-Cresol Purple | |
| 4-[3-(4-hydroxy-2-methylphenyl)-1,1-dioxo-2,1$l^{6}-benzoxathiol-3-yl]-3-methylphenol |
Specifications
| 2303-01-7 | |
| MFCD00005871 | |
| OLQIKGSZDTXODA-UHFFFAOYSA-N | |
| 4-[3-(4-hydroxy-2-methylphenyl)-1,1-dioxo-2,1$l^{6}-benzoxathiol-3-yl]-3-methylphenol | |
| 73030 | |
| 1 g | |
| Metacresol Purple |
| C21H18O5S | |
| m-Cresol Purple | |
| CC1=C(C=CC(=C1)O)C2(C3=CC=CC=C3S(=O)(=O)O2)C4=C(C=C(C=C4)O)C | |
| 382.43 | |
| 382.43 | |
| Crystal/Powder |
Safety and Handling
TSCA : Yes