missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Specifications
| Metacresol Purple | |
| C21H18O5S | |
| m-Cresol Purple | |
| CC1=C(C=CC(=C1)O)C2(C3=CC=CC=C3S(=O)(=O)O2)C4=C(C=C(C=C4)O)C | |
| 382.43 | |
| 382.43 | |
| Crystal/Powder |
| 2303-01-7 | |
| MFCD00005871 | |
| OLQIKGSZDTXODA-UHFFFAOYSA-N | |
| 4-[3-(4-hydroxy-2-methylphenyl)-1,1-dioxo-2,1$l^{6}-benzoxathiol-3-yl]-3-methylphenol | |
| 73030 | |
| 1 g |
Chemical Identifiers
| 2303-01-7 | |
| 382.43 | |
| OLQIKGSZDTXODA-UHFFFAOYSA-N | |
| 73030 | |
| CC1=C(C=CC(=C1)O)C2(C3=CC=CC=C3S(=O)(=O)O2)C4=C(C=C(C=C4)O)C |
| C21H18O5S | |
| MFCD00005871 | |
| m-Cresol Purple | |
| 4-[3-(4-hydroxy-2-methylphenyl)-1,1-dioxo-2,1$l^{6}-benzoxathiol-3-yl]-3-methylphenol |
Safety and Handling
TSCA : Yes