missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ MESP1 Monoclonal Antibody (1A3)

Mouse Monoclonal Antibody
Supplier: Invitrogen™ MA515645
Description
MA5-15645 targets MESP1 in indirect ELISA, WB applications and shows reactivity with Human samples. The MA5-15645 immunogen is purified recombinant fragment of human MESP1 expressed in E. Coli.. MA5-15645 detects MESP1 which has a predicted molecular weight of approximately 28.5kDa.
Mesp1 (mesoderm posterior 1) is a basic helix-loop-helix-type transcription factor, which along with its related proteins, has an essential role in both segmentation and rostro-caudal patterning of somites in the anterior pre-somitic mesoderm. Mesp1 is expressed in many of the precursors of the cardiovascular system and is known to play a role in the process of cardiac morphogenesis.Chemical Identifiers
| MESP1 | |
| Conc. Not Determined | |
| ELISA, Western Blot | |
| Unconjugated | |
| Mouse | |
| RUO | |
| Ascites with 0.03% sodium azide | |
| Q9BRJ9 | |
| 55897 | |
| Purified recombinant fragment of human MESP1 expressed in E. Coli. | |
| Antibody |
| 1A3 | |
| Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
| Monoclonal | |
| Liquid | |
| IgG1 | |
| Human | |
| MESP1 | |
| bHLHc5; Class C basic helix-loop-helix protein 5; mesoderm posterior 1; mesoderm posterior 1 homolog; mesoderm posterior basic helix-loop-helix transcription factor 1; mesoderm posterior bHLH transcription factor 1; mesoderm posterior protein 1; MESP1; MGC10676 | |
| MESP1 | |
| Primary |
Specifications
| MESP1 | |
| Monoclonal | |
| Conc. Not Determined | |
| Ascites with 0.03% sodium azide | |
| Q9BRJ9 | |
| MESP1 | |
| Purified recombinant fragment of human MESP1 expressed in E. Coli. | |
| RUO | |
| 55897 | |
| Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
| Liquid |
| ELISA, Western Blot | |
| 1A3 | |
| Unconjugated | |
| MESP1 | |
| bHLHc5; Class C basic helix-loop-helix protein 5; mesoderm posterior 1; mesoderm posterior 1 homolog; mesoderm posterior basic helix-loop-helix transcription factor 1; mesoderm posterior bHLH transcription factor 1; mesoderm posterior protein 1; MESP1; MGC10676 | |
| Mouse | |
| 100 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgG1 |