missing translation for 'onlineSavingsMsg'
Learn More

Mesotartaric Acid Monohydrate 90.0+%, TCI America™

Catalog No. M00705G
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
M00705G 5 g
M007025G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. M00705G Supplier TCI America Supplier No. M00705G

Chemical Identifiers

CAS 5990-63-6
Molecular Formula C4H8O7
Molecular Weight (g/mol) 168.101
MDL Number MFCD00063641
InChI Key UUDLQDCYDSATCH-NUGIMEKKSA-N
Synonym meso-Tartaric Acid
PubChem CID 16218804
IUPAC Name (2R,3S)-2,3-dihydroxybutanedioic acid;hydrate
SMILES C(C(C(=O)O)O)(C(=O)O)O.O

Specifications

Melting Point 75°C
Color White
Quantity 5 g
Formula Weight 150.09
Percent Purity ≥90.0% (HPLC)
Physical Form Crystalline Powder
Chemical Name or Material Mesotartaric Acid Monohydrate
EINECSNumber (2)-1456
TSCA Yes