missing translation for 'onlineSavingsMsg'
Learn More
Learn More
meso-Tetrakis(4-methoxyphenyl)porphine, 95%
CAS: 22112-78-3 | C48H38N4O4 | 734.856 g/mol
$117.22 - $589.62
Chemical Identifiers
| CAS | 22112-78-3 |
|---|---|
| Molecular Formula | C48H38N4O4 |
| Molecular Weight (g/mol) | 734.856 |
| MDL Number | MFCD09909566 |
| InChI Key | SKXWDGDNWBYACJ-UHFFFAOYSA-N |
| Synonym | 5,10,15,20-tetrakis 4-methoxyphenyl-21h,23h-porphine, 5,10,15,20-tetrakis 4-methoxyphenyl porphyrin, tetra p-methoxyphenyl porphyrin, 21h,23h-porphine, 5,10,15,20-tetrakis 4-methoxyphenyl, 5,10,15,20-tetrakis 4-methoxyphenyl-21,22-dihydroporphyrin, 2,7,12,17-tetrakis 4-methoxyphenyl-21,22,23,24-tetraazapentacyclo 16.2.1.1 3 ,?.1?, 1 1 .1 1 3 , 1 ? tetracosa-1,3,5,7,9,11 23 ,12,14,16,18 21 ,19-undecaene, 4-methoxy-1-7,12,17-tris 4-methoxyphenyl-21,22,23,24-tetraazapentacyclo 16.2 .1.1<3,6>.1<8,11>.1<13,16> tetracosa-1,3,5,7,9,11 23 ,12,14,16,18 21 ,19-undec aen-2-yl benzene, acmc-209fs6, tetrakis p-methoxyphenyl porphyrin, meso-tetra 4-methoxyphenyl porphine |
| PubChem CID | 140886 |
| IUPAC Name | 5,10,15,20-tetrakis(4-methoxyphenyl)-21,22-dihydroporphyrin |
| SMILES | COC1=CC=C(C=C1)C2=C3C=CC(=C(C4=CC=C(N4)C(=C5C=CC(=N5)C(=C6C=CC2=N6)C7=CC=C(C=C7)OC)C8=CC=C(C=C8)OC)C9=CC=C(C=C9)OC)N3 |
| Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
|---|---|---|---|---|---|---|---|---|---|
| Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
|
AAH2594603
|
Thermo Scientific Chemicals
H2594603 |
1 g |
Each for $117.22
|
|
|||||
|
AAH2594609
|
Thermo Scientific Chemicals
H2594609 |
10 g |
Each for $589.62
|
|
|||||
Description
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Chemical Identifiers
| 22112-78-3 | |
| 734.856 | |
| SKXWDGDNWBYACJ-UHFFFAOYSA-N | |
| 140886 | |
| COC1=CC=C(C=C1)C2=C3C=CC(=C(C4=CC=C(N4)C(=C5C=CC(=N5)C(=C6C=CC2=N6)C7=CC=C(C=C7)OC)C8=CC=C(C=C8)OC)C9=CC=C(C=C9)OC)N3 |
| C48H38N4O4 | |
| MFCD09909566 | |
| 5,10,15,20-tetrakis 4-methoxyphenyl-21h,23h-porphine, 5,10,15,20-tetrakis 4-methoxyphenyl porphyrin, tetra p-methoxyphenyl porphyrin, 21h,23h-porphine, 5,10,15,20-tetrakis 4-methoxyphenyl, 5,10,15,20-tetrakis 4-methoxyphenyl-21,22-dihydroporphyrin, 2,7,12,17-tetrakis 4-methoxyphenyl-21,22,23,24-tetraazapentacyclo 16.2.1.1 3 ,?.1?, 1 1 .1 1 3 , 1 ? tetracosa-1,3,5,7,9,11 23 ,12,14,16,18 21 ,19-undecaene, 4-methoxy-1-7,12,17-tris 4-methoxyphenyl-21,22,23,24-tetraazapentacyclo 16.2 .1.1<3,6>.1<8,11>.1<13,16> tetracosa-1,3,5,7,9,11 23 ,12,14,16,18 21 ,19-undec aen-2-yl benzene, acmc-209fs6, tetrakis p-methoxyphenyl porphyrin, meso-tetra 4-methoxyphenyl porphine | |
| 5,10,15,20-tetrakis(4-methoxyphenyl)-21,22-dihydroporphyrin |
Specifications
| 22112-78-3 | |
| MFCD09909566 | |
| 5,10,15,20-tetrakis 4-methoxyphenyl-21h,23h-porphine, 5,10,15,20-tetrakis 4-methoxyphenyl porphyrin, tetra p-methoxyphenyl porphyrin, 21h,23h-porphine, 5,10,15,20-tetrakis 4-methoxyphenyl, 5,10,15,20-tetrakis 4-methoxyphenyl-21,22-dihydroporphyrin, 2,7,12,17-tetrakis 4-methoxyphenyl-21,22,23,24-tetraazapentacyclo 16.2.1.1 3 ,?.1?, 1 1 .1 1 3 , 1 ? tetracosa-1,3,5,7,9,11 23 ,12,14,16,18 21 ,19-undecaene, 4-methoxy-1-7,12,17-tris 4-methoxyphenyl-21,22,23,24-tetraazapentacyclo 16.2 .1.1<3,6>.1<8,11>.1<13,16> tetracosa-1,3,5,7,9,11 23 ,12,14,16,18 21 ,19-undec aen-2-yl benzene, acmc-209fs6, tetrakis p-methoxyphenyl porphyrin, meso-tetra 4-methoxyphenyl porphine | |
| COC1=CC=C(C=C1)C2=C3C=CC(=C(C4=CC=C(N4)C(=C5C=CC(=N5)C(=C6C=CC2=N6)C7=CC=C(C=C7)OC)C8=CC=C(C=C8)OC)C9=CC=C(C=C9)OC)N3 | |
| 734.856 | |
| 734.87 | |
| meso-Tetrakis(4-methoxyphenyl)porphine |
| C48H38N4O4 | |
| 1 g | |
| SKXWDGDNWBYACJ-UHFFFAOYSA-N | |
| 5,10,15,20-tetrakis(4-methoxyphenyl)-21,22-dihydroporphyrin | |
| 140886 | |
| 95% |
Safety and Handling
EINECSNumber : 000-000-0
TSCA : No
Recommended Storage : Keep cold
RUO – Research Use Only