missing translation for 'onlineSavingsMsg'
Learn More

meso-Tetrakis(4-methoxyphenyl)porphine, 95%

CAS: 22112-78-3 | C48H38N4O4 | 734.856 g/mol

Supplier:  Thermo Scientific Chemicals H2594603

 View more versions of this product

Catalog No. AAH2594603


Only null left
Explore available promotions

Description

Description

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Specifications

Specifications

meso-Tetrakis(4-methoxyphenyl)porphine
C48H38N4O4
1 g
SKXWDGDNWBYACJ-UHFFFAOYSA-N
5,10,15,20-tetrakis(4-methoxyphenyl)-21,22-dihydroporphyrin
140886
95%
22112-78-3
MFCD09909566
5,10,15,20-tetrakis 4-methoxyphenyl-21h,23h-porphine, 5,10,15,20-tetrakis 4-methoxyphenyl porphyrin, tetra p-methoxyphenyl porphyrin, 21h,23h-porphine, 5,10,15,20-tetrakis 4-methoxyphenyl, 5,10,15,20-tetrakis 4-methoxyphenyl-21,22-dihydroporphyrin, 2,7,12,17-tetrakis 4-methoxyphenyl-21,22,23,24-tetraazapentacyclo 16.2.1.1 3 ,?.1?, 1 1 .1 1 3 , 1 ? tetracosa-1,3,5,7,9,11 23 ,12,14,16,18 21 ,19-undecaene, 4-methoxy-1-7,12,17-tris 4-methoxyphenyl-21,22,23,24-tetraazapentacyclo 16.2 .1.1<3,6>.1<8,11>.1<13,16> tetracosa-1,3,5,7,9,11 23 ,12,14,16,18 21 ,19-undec aen-2-yl benzene, acmc-209fs6, tetrakis p-methoxyphenyl porphyrin, meso-tetra 4-methoxyphenyl porphine
COC1=CC=C(C=C1)C2=C3C=CC(=C(C4=CC=C(N4)C(=C5C=CC(=N5)C(=C6C=CC2=N6)C7=CC=C(C=C7)OC)C8=CC=C(C=C8)OC)C9=CC=C(C=C9)OC)N3
734.856
734.87

Chemical Identifiers

22112-78-3
734.856
SKXWDGDNWBYACJ-UHFFFAOYSA-N
140886
COC1=CC=C(C=C1)C2=C3C=CC(=C(C4=CC=C(N4)C(=C5C=CC(=N5)C(=C6C=CC2=N6)C7=CC=C(C=C7)OC)C8=CC=C(C=C8)OC)C9=CC=C(C=C9)OC)N3
C48H38N4O4
MFCD09909566
5,10,15,20-tetrakis 4-methoxyphenyl-21h,23h-porphine, 5,10,15,20-tetrakis 4-methoxyphenyl porphyrin, tetra p-methoxyphenyl porphyrin, 21h,23h-porphine, 5,10,15,20-tetrakis 4-methoxyphenyl, 5,10,15,20-tetrakis 4-methoxyphenyl-21,22-dihydroporphyrin, 2,7,12,17-tetrakis 4-methoxyphenyl-21,22,23,24-tetraazapentacyclo 16.2.1.1 3 ,?.1?, 1 1 .1 1 3 , 1 ? tetracosa-1,3,5,7,9,11 23 ,12,14,16,18 21 ,19-undecaene, 4-methoxy-1-7,12,17-tris 4-methoxyphenyl-21,22,23,24-tetraazapentacyclo 16.2 .1.1<3,6>.1<8,11>.1<13,16> tetracosa-1,3,5,7,9,11 23 ,12,14,16,18 21 ,19-undec aen-2-yl benzene, acmc-209fs6, tetrakis p-methoxyphenyl porphyrin, meso-tetra 4-methoxyphenyl porphine
5,10,15,20-tetrakis(4-methoxyphenyl)-21,22-dihydroporphyrin
Product Suggestions

Product Suggestions

Videos
Safety and Handling

Safety and Handling

EINECSNumber : 000-000-0

TSCA : No

Recommended Storage : Keep cold

SDS
Documents

Documents

Promotions

Promotions

RUO – Research Use Only