missing translation for 'onlineSavingsMsg'
Learn More

meso-Erythritol 99.0+%, TCI America™

Catalog No. E002125G
Change view
Click to view available options
Quantity:
25 g
100 g
500 g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
E002125G 25 g
E0021100G 100 g
E0021500G 500 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. E002125G Supplier TCI America Supplier No. E002125G

Chemical Identifiers

CAS 149-32-6
Molecular Formula C4H10O4
Molecular Weight (g/mol) 122.12
MDL Number MFCD00004710
InChI Key UNXHWFMMPAWVPI-UHFFFAOYNA-N
Synonym erythritol, meso-erythritol, phycitol, erythrit, phycite, erythrol, 2r,3s-butane-1,2,3,4-tetrol, erythro-tetritol, mesoerythritol, erythrite
PubChem CID 222285
ChEBI CHEBI:17113
IUPAC Name butane-1,2,3,4-tetrol
SMILES OCC(O)C(O)CO

Specifications

Melting Point 120°C
Color White
Boiling Point 331°C
Quantity 25 g
Formula Weight 122.12
Percent Purity ≥99.0% (HPLC,T)
Physical Form Crystalline Powder
Chemical Name or Material meso-Erythritol
EINECSNumber (2)-3651
RTECSNumber KF2000000
TSCA Yes