missing translation for 'onlineSavingsMsg'
Learn More

meso-Erythritol, 99%

CAS: 149-32-6 | C4H10O4 | 122.12 g/mol

Supplier:  Thermo Scientific Chemicals A1581322

 View more versions of this product

Catalog No. AAA1581322


Only null left
Explore available promotions

Description

Description

meso-Erythritol is used to support the carbon source growth requirement of Gluconobacter bacteria. It is also used in the chemical analysis of the reactivity of various radicals and radical anions, in encapsulated phase change materials (PCMs) to study its potential use in microelectronics cooling applications, as a reference compound in purification and analytical procedures developed to test fermentation broths.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
meso-Erythritol is used to support the carbon source growth requirement of Gluconobacter bacteria. It is also used in the chemical analysis of the reactivity of various radicals and radical anions, in encapsulated phase change materials (PCMs) to study its potential use in microelectronics cooling applications, as a reference compound in purification and analytical procedures developed to test fermentation broths.

Solubility
Soluble in water (0.1 g/ml).

Notes
Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Store at -20°C.
Specifications

Specifications

meso-Erythritol
119°C to 124°C
99%
MFCD00004710
1719753
erythritol, meso-erythritol, phycitol, erythrit, phycite, erythrol, 2r,3s-butane-1,2,3,4-tetrol, erythro-tetritol, mesoerythritol, erythrite
UNXHWFMMPAWVPI-UHFFFAOYNA-N
butane-1,2,3,4-tetrol
222285
122.12
149-32-6
329°C to 331°C
C4H10O4
100 g
14,3675
Soluble in water (0.1g/ml).
OCC(O)C(O)CO
122.12
CHEBI:17113
99%

Chemical Identifiers

149-32-6
122.12
UNXHWFMMPAWVPI-UHFFFAOYNA-N
222285
butane-1,2,3,4-tetrol
C4H10O4
MFCD00004710
erythritol, meso-erythritol, phycitol, erythrit, phycite, erythrol, 2r,3s-butane-1,2,3,4-tetrol, erythro-tetritol, mesoerythritol, erythrite
CHEBI:17113
OCC(O)C(O)CO
Product Suggestions

Product Suggestions

Videos
Safety and Handling

Safety and Handling

EINECSNumber : 205-737-3

RTECSNumber : KF2000000

TSCA : Yes

Recommended Storage : Ambient temperatures

SDS
Documents

Documents

Promotions

Promotions

RUO – Research Use Only