missing translation for 'onlineSavingsMsg'
Learn More

meso-alpha,beta-Di(4-pyridyl) Glycol 98.0+%, TCI America™

Catalog No. D09365G
Change view
Click to view available options
Quantity:
5 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
D09365G 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. D09365G Supplier TCI America Supplier No. D09365G
Only null left

Chemical Identifiers

CAS 4972-49-0
Molecular Formula C12H12N2O2
Molecular Weight (g/mol) 216.24
MDL Number MFCD00023627
InChI Key DHKSJSQSVHHBPH-TXEJJXNPSA-N
Synonym meso-1,2-Bis(4-pyridyl) Glycol, meso-1,2-Di(4-pyridyl) Glycol, meso-1,2-Di(4-pyridyl)-1,2-ethanediol
PubChem CID 12785626
IUPAC Name (1R,2S)-1,2-dipyridin-4-ylethane-1,2-diol
SMILES C1=CN=CC=C1C(C(C2=CC=NC=C2)O)O

Specifications

Quantity 5 g
Formula Weight 216.24
Percent Purity ≥98.0% (HPLC,T)
Physical Form Crystalline Powder
Chemical Name or Material meso-alpha,beta-Di(4-pyridyl) Glycol