accessibility menu, dialog, popup

UserName

CAS RN 4972-49-0

NNHOOH

CAS RN 4972-49-0

IUPAC Name: 1,2-bis(pyridin-4-yl)ethane-1,2-diol
Synonyms: 1,2-bis(pyridin-4-yl)ethane-1,2-diol
Molecular Weight (g/mol): 216.24
Molecular Formula: C12H12N2O2
InChi Key: DHKSJSQSVHHBPH-UHFFFAOYNA-N
SMILES: OC(C(O)C1=CC=NC=C1)C1=CC=NC=C1
Percent Purity 
  • (1)
Physical Form 
  • (1)
Quantity 
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results