missing translation for 'onlineSavingsMsg'
Learn More

meso-2,3-Dimercaptosuccinic Acid 95.0+%, TCI America™

Catalog No. p-7126897
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
D17225G 5 g
D172225G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. D17225G Supplier TCI America Supplier No. D17225G
Only null left

Chemical Identifiers

CAS 304-55-2
Molecular Formula C4H6O4S2
Molecular Weight (g/mol) 182.208
MDL Number MFCD00064799
InChI Key ACTRVOBWPAIOHC-XIXRPRMCSA-N
Synonym succimer, meso-2,3-dimercaptosuccinic acid, meso-dimercaptosuccinic acid, dmsa, dim-sa, 2r,3s-rel-2,3-dimercaptosuccinic acid, succimero, succimerum, chemet, dimercaptosuccinic acid
PubChem CID 2724354
ChEBI CHEBI:63623
IUPAC Name (2S,3R)-2,3-bis(sulfanyl)butanedioic acid
SMILES C(C(C(=O)O)S)(C(=O)O)S

Specifications

Melting Point 198°C
Color White
Quantity 5 g
Formula Weight 182.21
Percent Purity ≥98.0% (T)
Physical Form Crystalline Powder
Chemical Name or Material meso-2,3-Dimercaptosuccinic Acid
RTECSNumber WM7650000
TSCA Yes