missing translation for 'onlineSavingsMsg'
Learn More
Learn More
meso-2,3-Dibromosuccinic Acid 98.0+%, TCI America™
$35.41 - $156.59
Chemical Identifiers
| CAS | 608-36-6 |
|---|---|
| Molecular Formula | C4H2Br2O4 |
| Molecular Weight (g/mol) | 273.87 |
| MDL Number | MFCD00066439 |
| InChI Key | FJWGRXKOBIVTFA-XIXRPRMCSA-L |
| Synonym | 2r,3s-2,3-dibromosuccinic acid, meso-2,3-dibromosuccinic acid, unii-d2ii9ugq9x, d2ii9ugq9x, butanedioic acid, 2,3-dibromo-, 2r,3s-rel, 2r,3s-rel-2,3-dibromosuccinic acid, 2r,3s-2,3-dibromobutanedioic acid, meso-dibromosuccinic acid, 2,3-dibromosuccinic acid, meso, 2,3-dibromosuccinic acid meso-form mi |
| PubChem CID | 641611 |
| IUPAC Name | (2R,3S)-2,3-dibromobutanedioate |
| SMILES | [O-]C(=O)[C@@H](Br)[C@@H](Br)C([O-])=O |
Chemical Identifiers
| 608-36-6 | |
| 273.87 | |
| FJWGRXKOBIVTFA-XIXRPRMCSA-L | |
| 641611 | |
| [O-]C(=O)[C@@H](Br)[C@@H](Br)C([O-])=O |
| C4H2Br2O4 | |
| MFCD00066439 | |
| 2r,3s-2,3-dibromosuccinic acid, meso-2,3-dibromosuccinic acid, unii-d2ii9ugq9x, d2ii9ugq9x, butanedioic acid, 2,3-dibromo-, 2r,3s-rel, 2r,3s-rel-2,3-dibromosuccinic acid, 2r,3s-2,3-dibromobutanedioic acid, meso-dibromosuccinic acid, 2,3-dibromosuccinic acid, meso, 2,3-dibromosuccinic acid meso-form mi | |
| (2R,3S)-2,3-dibromobutanedioate |
Specifications
| 608-36-6 | |
| White | |
| MFCD00066439 | |
| 3261 | |
| FJWGRXKOBIVTFA-XIXRPRMCSA-L | |
| (2R,3S)-2,3-dibromobutanedioate | |
| 641611 | |
| ≥98.0% (T) | |
| meso-2,3-Dibromosuccinic Acid |
| 256°C | |
| C4H2Br2O4 | |
| 25 g | |
| 2r,3s-2,3-dibromosuccinic acid, meso-2,3-dibromosuccinic acid, unii-d2ii9ugq9x, d2ii9ugq9x, butanedioic acid, 2,3-dibromo-, 2r,3s-rel, 2r,3s-rel-2,3-dibromosuccinic acid, 2r,3s-2,3-dibromobutanedioic acid, meso-dibromosuccinic acid, 2,3-dibromosuccinic acid, meso, 2,3-dibromosuccinic acid meso-form mi | |
| [O-]C(=O)[C@@H](Br)[C@@H](Br)C([O-])=O | |
| 273.87 | |
| 275.88 | |
| Crystalline Powder |
Safety and Handling
EINECSNumber : (2)-1167
TSCA : Yes