accessibility menu, dialog, popup

UserName

OO(S)Br(R)BrOO

CAS RN 608-36-6

IUPAC Name: (2R,3S)-2,3-dibromobutanedioate
Synonyms: (2R,3S)-2,3-dibromobutanedioate
Molecular Weight (g/mol): 273.87
Molecular Formula: C4H2Br2O4
InChi Key: FJWGRXKOBIVTFA-XIXRPRMCSA-L
SMILES: [O-]C(=O)[C@@H](Br)[C@@H](Br)C([O-])=O
Percent Purity 
  • (2)
  • (1)
Quantity 
  • (1)
  • (1)
  • (1)
Physical Form 
  • (2)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (1)
  • (2)
  • (1)
  • (2)
  • (1)
  • (1)
  • (1)
  • (1)
  • (2)