missing translation for 'onlineSavingsMsg'
Learn More

meso-2,3-Dibromosuccinic Acid 98.0+%, TCI America™

Catalog No. D0211500G
Change view
Click to view available options
Quantity:
25 g
500 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
D0211500G 500 g
D021125G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. D0211500G Supplier TCI America Supplier No. D0211500G
Only null left

Chemical Identifiers

CAS 608-36-6
Molecular Formula C4H2Br2O4
Molecular Weight (g/mol) 273.87
MDL Number MFCD00066439
InChI Key FJWGRXKOBIVTFA-XIXRPRMCSA-L
Synonym 2r,3s-2,3-dibromosuccinic acid, meso-2,3-dibromosuccinic acid, unii-d2ii9ugq9x, d2ii9ugq9x, butanedioic acid, 2,3-dibromo-, 2r,3s-rel, 2r,3s-rel-2,3-dibromosuccinic acid, 2r,3s-2,3-dibromobutanedioic acid, meso-dibromosuccinic acid, 2,3-dibromosuccinic acid, meso, 2,3-dibromosuccinic acid meso-form mi
PubChem CID 641611
IUPAC Name (2R,3S)-2,3-dibromobutanedioate
SMILES [O-]C(=O)[C@@H](Br)[C@@H](Br)C([O-])=O

Specifications

Melting Point 256°C
Color White
Quantity 500 g
UN Number 3261
Formula Weight 275.88
Percent Purity ≥98.0% (T)
Physical Form Crystalline Powder
Chemical Name or Material meso-2,3-Dibromosuccinic Acid
EINECSNumber (2)-1167
TSCA Yes