missing translation for 'onlineSavingsMsg'
Learn More
Learn More
meso-2,3-Dibromosuccinic Acid 98.0+%, TCI America™
Specifications
Specifications
| Melting Point | 256°C |
| Color | White |
| Quantity | 25 g |
| UN Number | 3261 |
| Formula Weight | 275.88 |
| Percent Purity | ≥98.0% (T) |
| Physical Form | Crystalline Powder |
| Chemical Name or Material | meso-2,3-Dibromosuccinic Acid |
Safety and Handling
| EINECSNumber | (2)-1167 |
| TSCA | Yes |