missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Mesitylenesulfonic acid sodium salt hemihydrate, 98%
CAS: 6148-75-0 | C9H11NaO3S | 222.23 g/mol
Supplier: Thermo Scientific Chemicals B2202922
Description
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Specifications
| Mesitylenesulfonic acid sodium salt hemihydrate | |
| >300°C | |
| MFCD00067712 | |
| 5784990 | |
| AOJUNZYQOYSGHT-UHFFFAOYSA-M | |
| sodium;2,4,6-trimethylbenzenesulfonate | |
| 23661859 | |
| 98% |
| 6148-75-0 | |
| C9H11NaO3S | |
| 100 g | |
| sodium mesitylenesulfonate, benzenesulfonic acid, 2,4,6-trimethyl-, sodium salt, sodium 2,4,6-trimethylbenzenesulfonate, sodium 2,4,6-trimethylbenzenesulphonate, mesitylenesulfonic acid sodium salt hemihydrate, mesitylenesulfonic acid sodium salt, benzenesulfonic acid, 2,4,6-trimethyl-, sodium salt 1:1, 2,4,6-trimethylbenzenesulfonic acid sodium salt, mesitylene sodium sulfonate, acmc-1bbae | |
| [Na+].CC1=CC(C)=C(C(C)=C1)S([O-])(=O)=O | |
| 222.23 | |
| 231.26 (222.25 Anhydrous) |
Chemical Identifiers
| 6148-75-0 | |
| 222.23 | |
| AOJUNZYQOYSGHT-UHFFFAOYSA-M | |
| 23661859 | |
| [Na+].CC1=CC(C)=C(C(C)=C1)S([O-])(=O)=O |
| C9H11NaO3S | |
| MFCD00067712 | |
| sodium mesitylenesulfonate, benzenesulfonic acid, 2,4,6-trimethyl-, sodium salt, sodium 2,4,6-trimethylbenzenesulfonate, sodium 2,4,6-trimethylbenzenesulphonate, mesitylenesulfonic acid sodium salt hemihydrate, mesitylenesulfonic acid sodium salt, benzenesulfonic acid, 2,4,6-trimethyl-, sodium salt 1:1, 2,4,6-trimethylbenzenesulfonic acid sodium salt, mesitylene sodium sulfonate, acmc-1bbae | |
| sodium;2,4,6-trimethylbenzenesulfonate |
Safety and Handling
EINECSNumber : 228-157-2
TSCA : Yes
Recommended Storage : Ambient temperatures
RUO – Research Use Only