missing translation for 'onlineSavingsMsg'
Learn More

Mesitylene, 97%, pure

CAS: 108-67-8 | C9H12 | 120.19 g/mol

$152.01 - $445.23

Chemical Identifiers

CAS 108-67-8
Molecular Formula C9H12
Molecular Weight (g/mol) 120.19
MDL Number MFCD00008538
InChI Key AUHZEENZYGFFBQ-UHFFFAOYSA-N
Synonym mesitylene, sym-trimethylbenzene, benzene, 1,3,5-trimethyl, fleet-x, trimethylbenzol, 3,5-dimethyltoluene, s-trimethylbenzene, hsdb 92, 2,4,6-trimethylbenzene, 1,3,5-trimethyl-benzene
PubChem CID 7947
ChEBI CHEBI:34833
IUPAC Name 1,3,5-trimethylbenzene
SMILES CC1=CC(=CC(=C1)C)C
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Packaging Price Quantity  
Catalog Number Mfr. No. Quantity Packaging Price Quantity  
AC161322500
SDS View Documents
Thermo Scientific Chemicals
161322500
250 mL Glass bottle
Each for $152.01
Only null left
 
AC161320010
SDS View Documents
Thermo Scientific Chemicals
161320010
1 L Glass bottle
Each for $445.23
Only null left
 
Description

Description

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Specifications

Chemical Identifiers

108-67-8
120.19
AUHZEENZYGFFBQ-UHFFFAOYSA-N
7947
1,3,5-trimethylbenzene
C9H12
MFCD00008538
mesitylene, sym-trimethylbenzene, benzene, 1,3,5-trimethyl, fleet-x, trimethylbenzol, 3,5-dimethyltoluene, s-trimethylbenzene, hsdb 92, 2,4,6-trimethylbenzene, 1,3,5-trimethyl-benzene
CHEBI:34833
CC1=CC(=CC(=C1)C)C

Specifications

108-67-8
Colorless
163.0°C to 166.0°C
Authentic
Glass bottle
1.4980 to 1.5000
MFCD00008538
05, 406
15, 5977
Solubility in water: 2.9g/L (20°C). Other solubilities: miscible with alcohol,benzene and ether
CC1=CC(=CC(=C1)C)C
120.19
0.834 mm2/s (20°C)
120.19
Pure
Mesitylene
-45.0°C
0.8675g/mL
44°C
96% min. (GC)
C9H12
C6H3(CH3)3
250 mL
0.8675
mesitylene, sym-trimethylbenzene, benzene, 1,3,5-trimethyl, fleet-x, trimethylbenzol, 3,5-dimethyltoluene, s-trimethylbenzene, hsdb 92, 2,4,6-trimethylbenzene, 1,3,5-trimethyl-benzene
AUHZEENZYGFFBQ-UHFFFAOYSA-N
1,3,5-trimethylbenzene
7947
CHEBI:34833
97%
Liquid
Videos
Safety and Handling

Safety and Handling

GHS H Statement
Flammable liquid and vapour.
May be fatal if swallowed and enters airways.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
May cause drowsiness or dizziness.<br

GHS P Statement
Keep away from heat/sparks/open flames/hot surfaces.
- No smoking.
IF ON SKIN: Wash with plenty of soap and water.
IF IN EYES: Rinse cautiously with water for several minutes.
Remove contact lenses,if present and ea

GHS Signal Word: Danger

EINECSNumber : 203-604-4

SDS
missing translation for 'documents'

missing translation for 'documents'

RUO – Research Use Only