missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Mesityl Iodide 98.0+%, TCI America™
Specifications
Specifications
| Melting Point | 31°C |
| Color | White |
| Boiling Point | 250°C |
| Quantity | 25 g |
| Formula Weight | 246.09 |
| Percent Purity | ≥98.0% (GC) |
| Physical Form | Crystalline Powder |
| Chemical Name or Material | Mesityl Iodide |
Safety and Handling
| TSCA | No |
| Recommended Storage | Refrigerator |